Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148991797-148992344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs555894537 Chr.5: 148992027-148992027 on Build GRCh38
rs58767790 Chr.5: 148992116-148992116 on Build GRCh38
rs34928648 Chr.5: 148992211-148992212 on Build GRCh38
rs955364767 Chr.5: 148992049-148992049 on Build GRCh38
rs536941130 Chr.5: 148992074-148992074 on Build GRCh38
rs537800973 Chr.5: 148992001-148992001 on Build GRCh38
rs370871430 Chr.5: 148991912-148991912 on Build GRCh38
rs560716887 Chr.5: 148992021-148992021 on Build GRCh38
rs906225345 Chr.5: 148991907-148991907 on Build GRCh38
rs541966418 Chr.5: 148992310-148992310 on Build GRCh38
rs1005760197 Chr.5: 148991934-148991934 on Build GRCh38
rs777584386 Chr.5: 148991922-148991922 on Build GRCh38
rs897335315 Chr.5: 148992309-148992309 on Build GRCh38
rs999796194 Chr.5: 148992175-148992175 on Build GRCh38
rs985449071 Chr.5: 148992058-148992058 on Build GRCh38
rs886060122 Chr.5: 148992242-148992242 on Build GRCh38
rs56848629 Chr.5: 148992269-148992269 on Build GRCh38
rs952655981 Chr.5: 148992023-148992023 on Build GRCh38
rs182516724 Chr.5: 148991871-148991871 on Build GRCh38
rs1019905533 Chr.5: 148991863-148991863 on Build GRCh38
rs970817007 Chr.5: 148991984-148991984 on Build GRCh38
rs1025506683 Chr.5: 148991942-148991942 on Build GRCh38
rs886060121 Chr.5: 148992058-148992058 on Build GRCh38
rs1010340871 Chr.5: 148991857-148991857 on Build GRCh38
rs911452905 Chr.5: 148992069-148992069 on Build GRCh38
rs555041878 Chr.5: 148991971-148991972 on Build GRCh38
rs137953180 Chr.5: 148992161-148992161 on Build GRCh38
rs564921950 Chr.5: 148992143-148992143 on Build GRCh38
rs894377852 Chr.5: 148992188-148992188 on Build GRCh38
rs574992954 Chr.5: 148991968-148991968 on Build GRCh38
rs569019867 Chr.5: 148992271-148992271 on Build GRCh38
rs375723413 Chr.5: 148991979-148991980 on Build GRCh38
rs961515933 Chr.5: 148991987-148991987 on Build GRCh38
rs1006207045 Chr.5: 148992242-148992242 on Build GRCh38
rs576816749 Chr.5: 148991885-148991885 on Build GRCh38
rs542030426 Chr.5: 148992018-148992018 on Build GRCh38
rs958807677 Chr.5: 148991923-148991923 on Build GRCh38
rs939055418 Chr.5: 148992179-148992179 on Build GRCh38
rs1024137777 Chr.5: 148992237-148992237 on Build GRCh38
rs904201004 Chr.5: 148992172-148992172 on Build GRCh38
rs922385106 Chr.5: 148992129-148992129 on Build GRCh38
rs1055314009 Chr.5: 148991918-148991918 on Build GRCh38
rs760916782 Chr.5: 148992155-148992155 on Build GRCh38
rs886060120 Chr.5: 148991959-148991959 on Build GRCh38
rs573227616 Chr.5: 148991853-148991853 on Build GRCh38
rs984092840 Chr.5: 148992292-148992292 on Build GRCh38
rs1054717339 Chr.5: 148992181-148992181 on Build GRCh38
rs772925748 Chr.5: 148992132-148992132 on Build GRCh38
rs910426200 Chr.5: 148991828-148991828 on Build GRCh38
rs150317192 Chr.5: 148991904-148991904 on Build GRCh38
rs942892553 Chr.5: 148992070-148992070 on Build GRCh38
rs534278887 Chr.5: 148991864-148991864 on Build GRCh38
rs943250289 Chr.5: 148991839-148991839 on Build GRCh38
rs767439910 Chr.5: 148992123-148992123 on Build GRCh38
rs550901114 Chr.5: 148992202-148992202 on Build GRCh38
rs1017502274 Chr.5: 148991976-148991976 on Build GRCh38
rs371274790 Chr.5: 148991876-148991876 on Build GRCh38
rs923079600 Chr.5: 148992205-148992205 on Build GRCh38
rs1014062042 Chr.5: 148991924-148991924 on Build GRCh38
rs544508111 Chr.5: 148991889-148991889 on Build GRCh38
rs374825054 Chr.5: 148991983-148991983 on Build GRCh38

View Full Product Details