The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003280.2 | NP_003271.1 | 1 - 2 | 49 | 97 |
GenBank mRNA | AK313743.1 | - | 1 - 2 | 49 | 97 |
BC030244.1 | - | 1 - 2 | 36 | 97 | |
X07897.1 | - | 1 - 2 | 49 | 97 |