The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281503.1 | NP_001268432.1 | 1 - 1 | 354 | 72 |
NM_052910.2 | NP_443142.1 | 1 - 1 | 354 | 72 | |
GenBank mRNA | AB067497.1 | - | 1 - 1 | 354 | 72 |
BC051738.1 | - | 1 - 1 | 255 | 72 | |
DC334447.1 | - | 1 - 1 | 170 | 72 |