The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083615.3 | NP_001077084.2 | 5 - 6 | 672 | 65 |
NM_138995.4 | NP_620482.3 | 5 - 6 | 672 | 65 | |
NR_045682.1 | 5 - 6 | 672 | 65 | ||
NR_045683.1 | 5 - 6 | 672 | 65 | ||
NR_045684.1 | 5 - 6 | 672 | 65 | ||
XM_006712299.3 | XP_006712362.1 | 5 - 6 | 560 | 65 | |
XM_011510654.2 | XP_011508956.1 | 5 - 6 | 560 | 65 | |
XM_011510655.2 | XP_011508957.1 | 5 - 6 | 560 | 65 | |
GenBank mRNA | AF369908.1 | - | 5 - 6 | 672 | 65 |
AF391554.1 | - | 5 - 6 | 672 | 65 | |
AF391555.1 | - | 5 - 6 | 672 | 65 | |
AF391556.1 | - | 5 - 6 | 672 | 65 | |
AF391557.1 | - | 5 - 6 | 672 | 65 | |
AF391558.1 | - | 5 - 6 | 672 | 65 | |
AK055778.1 | - | 5 - 6 | 560 | 65 | |
BC040019.1 | - | 4 - 5 | 527 | 65 | |
BC136620.1 | - | 5 - 6 | 609 | 65 | |
BC144307.1 | - | 5 - 6 | 609 | 65 |