The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015657.3 | NP_056472.2 | 1 - 2 | 271 | 77 |
NM_173076.2 | NP_775099.2 | 9 - 10 | 1286 | 77 | |
NR_103740.1 | 10 - 11 | 1310 | 77 | ||
XM_011510951.2 | XP_011509253.1 | 9 - 10 | 1449 | 77 | |
GenBank mRNA | AF418105.1 | - | 4 - 5 | 408 | 77 |
AY033486.1 | - | 1 - 2 | 271 | 77 | |
AY219711.1 | - | 9 - 10 | 1286 | 77 |