The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002263.3 | NP_002254.2 | 1 - 2 | 464 | 87 |
XM_011514585.1 | XP_011512887.1 | 1 - 2 | 170 | 87 | |
XM_011514587.2 | XP_011512889.1 | 1 - 2 | 170 | 87 | |
XM_017010836.1 | XP_016866325.1 | 1 - 2 | 170 | 87 | |
GenBank mRNA | AJ010479.1 | - | 1 - 2 | 177 | 87 |
AK297378.1 | - | 1 - 2 | 141 | 87 | |
AK303239.1 | - | 1 - 2 | 128 | 87 | |
BC000712.2 | - | 1 - 2 | 172 | 87 | |
BC063567.1 | - | 1 - 2 | 122 | 87 | |
BC073878.1 | - | 1 - 2 | 157 | 87 | |
BC098438.1 | - | 1 - 2 | 154 | 87 | |
BC121041.1 | - | 1 - 2 | 95 | 87 | |
BC121042.1 | - | 1 - 2 | 95 | 87 | |
BE885900.1 | - | 1 - 2 | 86 |