The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173566.2 | NP_775837.2 | 9 - 9 | 9257 | 113 |
XM_006724219.3 | XP_006724282.1 | 9 - 9 | 9817 | 113 | |
XM_006724220.3 | XP_006724283.1 | 9 - 9 | 9219 | 113 | |
XM_006724221.2 | XP_006724284.1 | 8 - 8 | 8773 | 113 | |
XM_011530080.2 | XP_011528382.1 | 10 - 10 | 10025 | 113 | |
XM_011530081.2 | XP_011528383.1 | 9 - 9 | 9901 | 113 | |
XM_017028736.1 | XP_016884225.1 | 9 - 9 | 10977 | 113 | |
XM_017028737.1 | XP_016884226.1 | 8 - 8 | 10415 | 113 | |
GenBank mRNA | AK055930.1 | - | 1 - 1 | 450 | 113 |
AK090566.1 | - | 1 - 1 | 1604 | 113 | |
BX647272.1 | - | 6 - 6 | 3559 | 113 |