The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080439.2 | NP_001073908.2 | 6 - 6 | 3426 | 124 |
XM_011524283.1 | XP_011522585.1 | 6 - 6 | 3439 | 124 | |
GenBank mRNA | AL137385.1 | - | 1 - 1 | 1453 | 124 |
BC033017.1 | - | 6 - 6 | 3390 | 124 | |
BC033020.1 | - | 6 - 6 | 3413 | 124 | |
BC115385.1 | - | 7 - 7 | 3002 | 124 | |
BC115386.1 | - | 7 - 7 | 2998 | 124 |