Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 14 - 15 1479 145
NM_145695.2 NP_663733.1 14 - 15 1479 145
XM_005249628.2 XP_005249685.1 15 - 16 2007 145
XM_005249629.2 XP_005249686.1 15 - 16 2004 145
XM_005249630.2 XP_005249687.1 14 - 15 1520 145
XM_005249631.3 XP_005249688.1 13 - 14 1735 145
XM_011515153.2 XP_011513455.1 15 - 16 1845 145
XM_011515154.2 XP_011513456.1 15 - 16 1561 145
XM_011515156.2 XP_011513458.1 14 - 15 1517 145
XM_011515157.2 XP_011513459.1 14 - 15 1524 145
XM_011515158.2 XP_011513460.1 13 - 14 1660 145
XM_017011783.1 XP_016867272.1 15 - 16 1898 145
XM_017011784.1 XP_016867273.1 15 - 16 1558 145
XM_017011785.1 XP_016867274.1 14 - 15 1792 145
XM_017011786.1 XP_016867275.1 14 - 15 1772 145
XM_017011787.1 XP_016867276.1 14 - 15 1522 145
XM_017011788.1 XP_016867277.1 14 - 15 1808 145
XM_017011789.1 XP_016867278.1 14 - 15 1525 145
XM_017011790.1 XP_016867279.1 15 - 16 1844 145
XM_017011791.1 XP_016867280.1 15 - 16 1843 145
XM_017011792.1 XP_016867281.1 15 - 16 1843 145
GenBank mRNA AB018261.1 - 11 - 12 1108 145
AK304719.1 - 13 - 14 1735 145
BC105005.1 - 14 - 15 1359 145
BC143632.1 - 14 - 15 1356 145
BC143633.1 - 13 - 14 1323 145

View Full Product Details