Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location
Chr.11: 47465937 - 47565569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011519852.2 XP_011518154.1 15 - 15 2000 88
XM_011519854.1 XP_011518156.1 17 - 17 2108 88
XM_011519855.1 XP_011518157.1 13 - 13 1829 88
XM_011519856.1 XP_011518158.1 14 - 14 1910 88
XM_011519857.1 XP_011518159.1 13 - 13 1781 88
XM_011519858.1 XP_011518160.1 13 - 13 1876 88
XM_017017101.1 XP_016872590.1 16 - 16 1992 88
XM_017017108.1 XP_016872597.1 14 - 14 1792 88
XM_017017111.1 XP_016872600.1 14 - 14 1789 88
XM_017017115.1 XP_016872604.1 15 - 15 1832 88
XM_017017116.1 XP_016872605.1 14 - 14 1867 88
XM_017017117.1 XP_016872606.1 13 - 13 1628 88
XM_017017124.1 XP_016872613.1 15 - 15 1920 88
XM_017017125.1 XP_016872614.1 13 - 13 1649 88
XM_017017126.1 XP_016872615.1 13 - 13 1778 88
XM_017017127.1 XP_016872616.1 13 - 13 1802 88
XM_017017133.1 XP_016872622.1 13 - 13 1608 88
XM_017017135.1 XP_016872624.1 13 - 13 1766 88
GenBank mRNA AF248648.1 - 12 - 12 1585 88
AK056024.1 - 1 - 1 152 88

View Full Product Details