Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
Chromosome Location
Chr.1: 45620044 - 45624787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017000451.1 XP_016855940.1 1 - 2 1056 80
XM_017000454.1 XP_016855943.1 1 - 2 322 80
GenBank mRNA AK057646.1 - 1 - 2 327 80

View Full Product Details