Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16989744-16990236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607272 c.2363C>T p.P788L Chr.1: 16990161-16990161 on Build GRCh38
5480937 c.2309A>G p.Q770R Chr.1: 16990230-16990230 on Build GRCh38
1245700 c.2379G>T p.P793P Chr.1: 16990160-16990160 on Build GRCh38
7356922 c.2413G>A p.D805N Chr.1: 16990003-16990003 on Build GRCh38
3802889 c.2368G>A p.E790K Chr.1: 16990171-16990171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761491667 Chr.1: 16990039-16990039 on Build GRCh38
rs766432992 Chr.1: 16989946-16989946 on Build GRCh38
rs199655251 Chr.1: 16990147-16990147 on Build GRCh38
rs761620417 Chr.1: 16989935-16989935 on Build GRCh38
rs56170027 Chr.1: 16990213-16990213 on Build GRCh38
rs759587904 Chr.1: 16990157-16990157 on Build GRCh38
rs755936102 Chr.1: 16990098-16990100 on Build GRCh38
rs144708504 Chr.1: 16990208-16990208 on Build GRCh38
rs986226126 Chr.1: 16989819-16989819 on Build GRCh38
rs763659305 Chr.1: 16989884-16989884 on Build GRCh38
rs77880760 Chr.1: 16990082-16990082 on Build GRCh38
rs147611385 Chr.1: 16989938-16989938 on Build GRCh38
rs765064192 Chr.1: 16989937-16989937 on Build GRCh38
rs978493820 Chr.1: 16990127-16990127 on Build GRCh38
rs537112220 Chr.1: 16989871-16989871 on Build GRCh38
rs769392537 Chr.1: 16990199-16990199 on Build GRCh38
rs954971480 Chr.1: 16989800-16989800 on Build GRCh38
rs1030482567 Chr.1: 16990050-16990050 on Build GRCh38
rs949240567 Chr.1: 16989810-16989810 on Build GRCh38
rs760318841 Chr.1: 16989882-16989882 on Build GRCh38
rs757153866 Chr.1: 16989841-16989841 on Build GRCh38
rs757799277 Chr.1: 16990109-16990109 on Build GRCh38
rs752487771 Chr.1: 16990135-16990135 on Build GRCh38
rs768017868 Chr.1: 16990121-16990121 on Build GRCh38
rs372376094 Chr.1: 16989965-16989965 on Build GRCh38
rs780146972 Chr.1: 16989854-16989854 on Build GRCh38
rs200809857 Chr.1: 16989878-16989878 on Build GRCh38
rs778229092 Chr.1: 16990087-16990087 on Build GRCh38
rs917176878 Chr.1: 16989891-16989891 on Build GRCh38
rs746438512 Chr.1: 16990115-16990115 on Build GRCh38
rs376237408 Chr.1: 16989955-16989955 on Build GRCh38
rs755157804 Chr.1: 16989957-16989957 on Build GRCh38
rs748463334 Chr.1: 16989869-16989869 on Build GRCh38
rs1037763231 Chr.1: 16990014-16990014 on Build GRCh38
rs766227206 Chr.1: 16990077-16990077 on Build GRCh38
rs747248855 Chr.1: 16989856-16989856 on Build GRCh38
rs778353567 Chr.1: 16989979-16989979 on Build GRCh38
rs148374514 Chr.1: 16990214-16990214 on Build GRCh38
rs867014635 Chr.1: 16990200-16990200 on Build GRCh38
rs777422289 Chr.1: 16990206-16990206 on Build GRCh38
rs897837480 Chr.1: 16989848-16989848 on Build GRCh38
rs535209471 Chr.1: 16989821-16989821 on Build GRCh38
rs771708290 Chr.1: 16989879-16989879 on Build GRCh38
rs374203880 Chr.1: 16990097-16990097 on Build GRCh38
rs770849259 Chr.1: 16990148-16990148 on Build GRCh38
rs779312019 Chr.1: 16990110-16990110 on Build GRCh38
rs776124539 Chr.1: 16990033-16990033 on Build GRCh38
rs866035312 Chr.1: 16989961-16989961 on Build GRCh38
rs751615609 Chr.1: 16990081-16990081 on Build GRCh38
rs763937012 Chr.1: 16989820-16989820 on Build GRCh38
rs781557299 Chr.1: 16989861-16989861 on Build GRCh38
rs747785443 Chr.1: 16990132-16990132 on Build GRCh38
rs773837281 Chr.1: 16989877-16989877 on Build GRCh38
rs754208001 Chr.1: 16990171-16990171 on Build GRCh38
rs765011219 Chr.1: 16990040-16990040 on Build GRCh38
rs140048110 Chr.1: 16990172-16990172 on Build GRCh38
rs376236999 Chr.1: 16990192-16990192 on Build GRCh38
rs941921003 Chr.1: 16990066-16990066 on Build GRCh38
rs137955309 Chr.1: 16990191-16990191 on Build GRCh38
rs774997717 Chr.1: 16989994-16989994 on Build GRCh38
rs1017806715 Chr.1: 16990211-16990211 on Build GRCh38
rs374008623 Chr.1: 16989779-16989779 on Build GRCh38
rs55929813 Chr.1: 16989826-16989826 on Build GRCh38
rs764444063 Chr.1: 16990163-16990163 on Build GRCh38
rs759142662 Chr.1: 16989784-16989784 on Build GRCh38
rs772920644 Chr.1: 16990142-16990142 on Build GRCh38
rs994103230 Chr.1: 16990035-16990035 on Build GRCh38
rs368244226 Chr.1: 16989977-16989977 on Build GRCh38
rs868725919 Chr.1: 16989764-16989764 on Build GRCh38
rs768307347 Chr.1: 16990004-16990004 on Build GRCh38
rs781298587 Chr.1: 16989960-16989960 on Build GRCh38
rs773047666 Chr.1: 16990044-16990044 on Build GRCh38
rs756467036 Chr.1: 16990085-16990085 on Build GRCh38
rs778854959 Chr.1: 16989846-16989846 on Build GRCh38
rs377073826 Chr.1: 16989851-16989851 on Build GRCh38
rs752965372 Chr.1: 16990084-16990084 on Build GRCh38
rs541344554 Chr.1: 16990046-16990046 on Build GRCh38
rs749802732 Chr.1: 16989987-16989987 on Build GRCh38
rs756052051 Chr.1: 16990053-16990053 on Build GRCh38
rs760315801 Chr.1: 16989816-16989816 on Build GRCh38
rs746533808 Chr.1: 16990003-16990003 on Build GRCh38
rs771579345 Chr.1: 16989992-16989992 on Build GRCh38
rs375894941 Chr.1: 16989773-16989773 on Build GRCh38
rs886045580 Chr.1: 16990133-16990133 on Build GRCh38
rs750889358 Chr.1: 16990182-16990182 on Build GRCh38
rs151222153 Chr.1: 16990158-16990158 on Build GRCh38
rs1017231900 Chr.1: 16990083-16990083 on Build GRCh38
rs182748624 Chr.1: 16990160-16990160 on Build GRCh38
rs758449038 Chr.1: 16989853-16989853 on Build GRCh38
rs774130460 Chr.1: 16990154-16990154 on Build GRCh38
rs371083479 Chr.1: 16989860-16989860 on Build GRCh38
rs201321612 Chr.1: 16989825-16989825 on Build GRCh38
rs376087842 Chr.1: 16990052-16990052 on Build GRCh38
rs898009726 Chr.1: 16990017-16990017 on Build GRCh38
rs779373993 Chr.1: 16990180-16990180 on Build GRCh38
rs748454159 Chr.1: 16989976-16989976 on Build GRCh38
rs770434494 Chr.1: 16989776-16989776 on Build GRCh38
rs752367474 Chr.1: 16989795-16989795 on Build GRCh38
rs767492622 Chr.1: 16990079-16990086 on Build GRCh38
rs760914179 Chr.1: 16990161-16990161 on Build GRCh38
rs779911360 Chr.1: 16990102-16990113 on Build GRCh38
rs770381718 Chr.1: 16989876-16989876 on Build GRCh38
rs776925680 Chr.1: 16989765-16989765 on Build GRCh38
rs774924482 Chr.1: 16989881-16989881 on Build GRCh38
rs780819882 Chr.1: 16990124-16990124 on Build GRCh38
rs776448394 Chr.1: 16989886-16989886 on Build GRCh38
rs986327732 Chr.1: 16989868-16989868 on Build GRCh38
rs905157690 Chr.1: 16989980-16989980 on Build GRCh38
rs748986186 Chr.1: 16990207-16990207 on Build GRCh38
rs935118855 Chr.1: 16989782-16989782 on Build GRCh38
rs750369195 Chr.1: 16990090-16990094 on Build GRCh38

View Full Product Details