Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13921930-13922440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629779 c.319A>T p.I107L Chr.16: 13922142-13922142 on Build GRCh38
6944816 c.329C>T p.T110I Chr.16: 13922152-13922152 on Build GRCh38
1629778 c.307C>G p.Q103E Chr.16: 13922130-13922130 on Build GRCh38
6838968 c.357G>T p.L119F Chr.16: 13922180-13922180 on Build GRCh38
3361637 c.209A>G p.E70G Chr.16: 13922032-13922032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761146709 Chr.16: 13922253-13922254 on Build GRCh38
rs771203473 Chr.16: 13922057-13922059 on Build GRCh38
rs555020322 Chr.16: 13922186-13922186 on Build GRCh38
rs577056376 Chr.16: 13922384-13922384 on Build GRCh38
rs563647036 Chr.16: 13922387-13922387 on Build GRCh38
rs55761944 Chr.16: 13922064-13922064 on Build GRCh38
rs540569071 Chr.16: 13922307-13922307 on Build GRCh38
rs763208431 Chr.16: 13922137-13922137 on Build GRCh38
rs941012957 Chr.16: 13922200-13922200 on Build GRCh38
rs955842806 Chr.16: 13922374-13922374 on Build GRCh38
rs752008977 Chr.16: 13922142-13922142 on Build GRCh38
rs192488568 Chr.16: 13922048-13922048 on Build GRCh38
rs371487368 Chr.16: 13922083-13922083 on Build GRCh38
rs556330628 Chr.16: 13922098-13922098 on Build GRCh38
rs773652331 Chr.16: 13922131-13922131 on Build GRCh38
rs953913935 Chr.16: 13922336-13922336 on Build GRCh38
rs144666685 Chr.16: 13922190-13922190 on Build GRCh38
rs763811136 Chr.16: 13922169-13922169 on Build GRCh38
rs1028025892 Chr.16: 13922326-13922326 on Build GRCh38
rs771860466 Chr.16: 13922033-13922033 on Build GRCh38
rs141591400 Chr.16: 13922040-13922040 on Build GRCh38
rs777834718 Chr.16: 13922183-13922183 on Build GRCh38
rs896008461 Chr.16: 13922341-13922341 on Build GRCh38
rs780096152 Chr.16: 13922195-13922195 on Build GRCh38
rs3136057 Chr.16: 13922338-13922338 on Build GRCh38
rs912317139 Chr.16: 13922364-13922364 on Build GRCh38
rs918305286 Chr.16: 13922227-13922227 on Build GRCh38
rs747613537 Chr.16: 13922239-13922239 on Build GRCh38
rs767586458 Chr.16: 13922149-13922149 on Build GRCh38
rs772418764 Chr.16: 13922252-13922253 on Build GRCh38
rs187435008 Chr.16: 13922080-13922080 on Build GRCh38
rs747024254 Chr.16: 13922001-13922001 on Build GRCh38
rs753534967 Chr.16: 13922049-13922049 on Build GRCh38
rs565715127 Chr.16: 13922178-13922178 on Build GRCh38
rs774988274 Chr.16: 13922228-13922233 on Build GRCh38
rs768893462 Chr.16: 13922009-13922009 on Build GRCh38
rs145315496 Chr.16: 13922034-13922034 on Build GRCh38
rs34861763 Chr.16: 13922264-13922265 on Build GRCh38
rs776312000 Chr.16: 13922237-13922237 on Build GRCh38
rs746957529 Chr.16: 13922230-13922230 on Build GRCh38
rs755021656 Chr.16: 13922070-13922070 on Build GRCh38
rs144408815 Chr.16: 13922128-13922128 on Build GRCh38
rs61760162 Chr.16: 13922051-13922051 on Build GRCh38
rs760327512 Chr.16: 13922112-13922112 on Build GRCh38
rs140822771 Chr.16: 13922301-13922302 on Build GRCh38
rs373361309 Chr.16: 13922231-13922236 on Build GRCh38
rs768649885 Chr.16: 13922231-13922231 on Build GRCh38
rs769932063 Chr.16: 13922079-13922079 on Build GRCh38
rs763620308 Chr.16: 13922253-13922255 on Build GRCh38
rs759518166 Chr.16: 13922031-13922031 on Build GRCh38
rs746393376 Chr.16: 13922232-13922233 on Build GRCh38
rs1015821969 Chr.16: 13922265-13922265 on Build GRCh38
rs1014546174 Chr.16: 13922071-13922071 on Build GRCh38
rs750687183 Chr.16: 13922062-13922062 on Build GRCh38
rs1014905315 Chr.16: 13922358-13922358 on Build GRCh38
rs769539825 Chr.16: 13922252-13922252 on Build GRCh38
rs749125796 Chr.16: 13922015-13922015 on Build GRCh38
rs139142361 Chr.16: 13921997-13921997 on Build GRCh38
rs549297996 Chr.16: 13922144-13922144 on Build GRCh38
rs772713662 Chr.16: 13922258-13922258 on Build GRCh38
rs189742296 Chr.16: 13922174-13922174 on Build GRCh38
rs200805983 Chr.16: 13921992-13921992 on Build GRCh38
rs766951888 Chr.16: 13922140-13922140 on Build GRCh38
rs763445140 Chr.16: 13922263-13922263 on Build GRCh38
rs775257742 Chr.16: 13922109-13922109 on Build GRCh38
rs750930128 Chr.16: 13921987-13921987 on Build GRCh38
rs776473010 Chr.16: 13922396-13922396 on Build GRCh38
rs751713894 Chr.16: 13922068-13922068 on Build GRCh38
rs764943215 Chr.16: 13922129-13922129 on Build GRCh38
rs3136056 Chr.16: 13922075-13922075 on Build GRCh38
rs113434368 Chr.16: 13922253-13922253 on Build GRCh38
rs985418503 Chr.16: 13922175-13922175 on Build GRCh38
rs377085353 Chr.16: 13922150-13922150 on Build GRCh38
rs3136058 Chr.16: 13922355-13922355 on Build GRCh38
rs191886782 Chr.16: 13922082-13922082 on Build GRCh38
rs770696743 Chr.16: 13922016-13922016 on Build GRCh38
rs1032984252 Chr.16: 13922397-13922397 on Build GRCh38
rs1035229051 Chr.16: 13922042-13922042 on Build GRCh38
rs575078496 Chr.16: 13922278-13922278 on Build GRCh38
rs748380872 Chr.16: 13922076-13922076 on Build GRCh38
rs773956647 Chr.16: 13922028-13922028 on Build GRCh38
rs768270013 Chr.16: 13922113-13922113 on Build GRCh38
rs781373518 Chr.16: 13922012-13922012 on Build GRCh38
rs1038044252 Chr.16: 13922226-13922226 on Build GRCh38
rs986588983 Chr.16: 13922346-13922346 on Build GRCh38
rs148791570 Chr.16: 13922148-13922148 on Build GRCh38
rs765712858 Chr.16: 13922055-13922055 on Build GRCh38

View Full Product Details