Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA5A
Entrez Gene ID
763
Gene Name
carbonic anhydrase 5A
Gene Aliases
CA5, CA5AD, CAV, CAVA, GS1-21A4.1
Chromosome Location
Chr.16: 87888019 - 87936575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001739.1 NP_001730.1 1 - 2 195 107
XM_005256134.4 XP_005256191.1 1 - 2 264 107
XM_011523309.2 XP_011521611.1 1 - 2 264 107
XM_011523310.2 XP_011521612.1 1 - 2 264 107
XM_017023646.1 XP_016879135.1 1 - 2 264 107
GenBank mRNA BC137405.1 - 1 - 2 191 107
BC137411.1 - 1 - 2 191 107
L19297.1 - 1 - 2 194 107

View Full Product Details