Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
Chromosome Location
Chr.15: 75355792 - 75368680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256494.1 NP_001243423.1 3 - 4 396 57
NM_001256495.1 NP_001243424.1 3 - 4 396 57
NM_001256496.1 NP_001243425.1 3 - 4 396 57
NM_006715.3 NP_006706.2 3 - 4 396 57
XM_005254384.2 XP_005254441.1 3 - 4 449 57
XM_017022186.1 XP_016877675.1 2 - 3 744 57
XM_017022187.1 XP_016877676.1 2 - 3 744 57
XM_017022188.1 XP_016877677.1 3 - 4 449 57
GenBank mRNA AB209141.1 - - 274 57
AF044414.2 - 3 - 4 408 57
AK225145.1 - 3 - 4 373 57
AK294892.1 - 3 - 4 375 57
AK295265.1 - 3 - 4 705 57
AK311065.1 - 2 - 3 248 57
AL136876.1 - 3 - 4 357 57
BC050550.1 - 3 - 4 363 57
BC080191.1 - 3 - 4 361 57
DC342919.1 - 3 - 4 399 57

View Full Product Details