Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
Chromosome Location
Chr.7: 100177563 - 100219334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282716.1 NP_001269645.1 18 - 19 2105 71
NM_001282717.1 NP_001269646.1 18 - 19 2186 71
NM_001282718.1 NP_001269647.1 16 - 17 1979 71
NM_012447.3 NP_036579.2 18 - 19 2186 71
XM_011515742.1 XP_011514044.1 18 - 19 2153 71
XM_017011683.1 XP_016867172.1 18 - 19 2033 71
XM_017011684.1 XP_016867173.1 18 - 19 2153 71
XM_017011685.1 XP_016867174.1 18 - 19 2266 71
XM_017011686.1 XP_016867175.1 18 - 19 2033 71
XM_017011687.1 XP_016867176.1 16 - 17 1859 71
GenBank mRNA AJ007798.2 - 18 - 19 2106 71
AK292394.1 - 18 - 19 2013 71
AK302587.1 - 15 - 16 1597 71
AK302693.1 - 16 - 17 1802 71
AL833816.1 - 17 - 18 2277 71
BC140932.1 - 18 - 19 1945 71
BC146806.1 - 18 - 19 2011 71

View Full Product Details