Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFYC
Entrez Gene ID
4802
Gene Name
nuclear transcription factor Y subunit gamma
Gene Aliases
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
Chromosome Location
Chr.1: 40691570 - 40771603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142587.1 NP_001136059.1 4 - 5 595 128
NM_001142588.1 NP_001136060.1 4 - 5 595 128
NM_001142590.1 NP_001136062.1 9 - 10 972 128
NM_001308114.1 NP_001295043.1 3 - 4 297 128
NM_001308115.1 NP_001295044.1 4 - 5 591 128
NM_014223.4 NP_055038.2 4 - 5 595 128
XM_005270894.1 XP_005270951.1 4 - 5 591 128
XM_005270895.3 XP_005270952.3 4 - 5 447 128
XM_006710658.2 XP_006710721.2 4 - 5 447 128
XM_006710660.2 XP_006710723.2 4 - 5 447 128
XM_006710661.1 XP_006710724.1 4 - 5 476 128
XM_006710662.2 XP_006710725.2 4 - 5 447 128
XM_011541516.1 XP_011539818.1 4 - 5 396 128
XM_011541517.1 XP_011539819.1 4 - 5 447 128
XM_017001364.1 XP_016856853.1 4 - 5 616 128
XM_017001365.1 XP_016856854.1 4 - 5 476 128
XM_017001366.1 XP_016856855.1 4 - 5 550 128
XM_017001367.1 XP_016856856.1 4 - 5 591 128
XM_017001368.1 XP_016856857.1 4 - 5 591 128
XM_017001369.1 XP_016856858.1 5 - 6 948 128
GenBank mRNA AB208975.1 - 4 - 5 422 128
AF191744.1 - 3 - 4 296 128
AK000346.1 - 4 - 5 548 128
AK127677.1 - 9 - 10 967 128
AK300774.1 - 4 - 5 398 128
BC005003.1 - 4 - 5 444 128
BT020081.1 - 3 - 4 289 128
D85425.1 - 4 - 5 372 128
U62296.1 - 3 - 4 289 128
U78774.1 - 5 - 6 463 128

View Full Product Details