Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
Chromosome Location
Chr.X: 108155607 - 108439497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287758.1 NP_001274687.1 4 - 5 515 64
NM_001287759.1 NP_001274688.1 4 - 5 515 64
NM_001287760.1 NP_001274689.1 4 - 5 515 64
NM_001847.3 NP_001838.2 4 - 5 410 64
NM_033641.3 NP_378667.1 4 - 5 515 64
XM_006724617.3 XP_006724680.1 4 - 5 404 64
XM_011530852.2 XP_011529154.1 4 - 5 404 64
XM_011530853.2 XP_011529155.1 2 - 3 1120 64
XM_011530854.2 XP_011529156.1 4 - 5 405 64
GenBank mRNA BC000874.1 - 4 - 5 449 64
BC140855.1 - 4 - 5 411 64
BC144669.1 - 4 - 5 411 64
CN313347.1 - 4 - 5 518 64
D21337.1 - 4 - 5 515 64
L22763.1 - 4 - 5 393 64
U04845.1 - 4 - 5 393 64

View Full Product Details