The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001897.4 | NP_001888.2 | 4 - 5 | 4368 | 60 |
GenBank mRNA | AY359468.1 | - | 4 - 5 | 4275 | 60 |
X96753.1 | - | 4 - 5 | 4275 | 60 |