Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1G
Entrez Gene ID
8913
Gene Name
calcium voltage-gated channel subunit alpha1 G
Gene Aliases
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location
Chr.17: 50560929 - 50629625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256324.1 NP_001243253.1 4 - 5 979 57
NM_001256325.1 NP_001243254.1 4 - 5 979 57
NM_001256326.1 NP_001243255.1 4 - 5 979 57
NM_001256327.1 NP_001243256.1 4 - 5 979 57
NM_001256328.1 NP_001243257.1 4 - 5 979 57
NM_001256329.1 NP_001243258.1 4 - 5 979 57
NM_001256330.1 NP_001243259.1 4 - 5 979 57
NM_001256331.1 NP_001243260.1 4 - 5 979 57
NM_001256332.1 NP_001243261.1 4 - 5 979 57
NM_001256333.1 NP_001243262.1 4 - 5 979 57
NM_001256334.1 NP_001243263.1 4 - 5 979 57
NM_001256359.1 NP_001243288.1 4 - 5 979 57
NM_001256360.1 NP_001243289.1 4 - 5 979 57
NM_001256361.1 NP_001243290.1 4 - 5 979 57
NM_018896.4 NP_061496.2 4 - 5 979 57
NM_198376.2 NP_938190.1 4 - 5 979 57
NM_198377.2 NP_938191.2 4 - 5 979 57
NM_198378.2 NP_938192.1 4 - 5 979 57
NM_198379.2 NP_938193.1 4 - 5 979 57
NM_198380.2 NP_938194.1 4 - 5 979 57
NM_198382.2 NP_938196.1 4 - 5 979 57
NM_198383.2 NP_938197.1 4 - 5 979 57
NM_198384.2 NP_938198.1 4 - 5 979 57
NM_198385.2 NP_938199.1 4 - 5 979 57
NM_198386.2 NP_938200.1 4 - 5 979 57
NM_198387.2 NP_938201.1 4 - 5 979 57
NM_198388.2 NP_938202.1 4 - 5 979 57
NM_198396.2 NP_938406.1 4 - 5 979 57
NR_046054.1 4 - 5 979 57
NR_046055.1 4 - 5 979 57
NR_046056.1 4 - 5 979 57
NR_046057.1 4 - 5 979 57
NR_046058.1 4 - 5 979 57
XM_006722160.3 XP_006722223.1 4 - 5 1118 57
XM_006722161.3 XP_006722224.1 4 - 5 1118 57
GenBank mRNA AB012043.1 - 4 - 5 615 57
AF124351.1 - 4 - 5 959 57
AF126965.1 - 4 - 5 762 57
AF126966.1 - 4 - 5 762 57
AF134985.1 - 4 - 5 587 57
AF134986.1 - 4 - 5 587 57
AF190860.1 - 4 - 5 803 57
AF227744.1 - 4 - 5 587 57
AF227745.1 - 4 - 5 587 57
AF227746.1 - 4 - 5 587 57
AF227747.1 - 4 - 5 587 57
AF227748.1 - 4 - 5 587 57
AF227749.1 - 4 - 5 587 57
AF227750.1 - 4 - 5 587 57
AF227751.1 - 4 - 5 587 57
BC110995.1 - 4 - 5 979 57
DQ494449.1 - 4 - 5 587 57
DQ494450.1 - 4 - 5 587 57
DQ494451.1 - 4 - 5 587 57
DQ494452.1 - 4 - 5 587 57
DQ494453.1 - 4 - 5 587 57
DQ494454.1 - 4 - 5 587 57
DQ494455.1 - 4 - 5 587 57
DQ494456.1 - 4 - 5 587 57
DQ494457.1 - 4 - 5 587 57
DQ494458.1 - 4 - 5 587 57
DQ494459.1 - 4 - 5 587 57
DQ494460.1 - 4 - 5 587 57
DQ494461.1 - 4 - 5 587 57
DQ494462.1 - 4 - 5 587 57
DQ494463.1 - 4 - 5 587 57
DQ494464.1 - 4 - 5 587 57
DQ494465.1 - 4 - 5 587 57
DQ494466.1 - 4 - 5 587 57
DQ494467.1 - 4 - 5 587 57
DQ494468.1 - 4 - 5 587 57
DQ494469.1 - 4 - 5 587 57
DQ494470.1 - 4 - 5 587 57
DQ494471.1 - 4 - 5 587 57
DQ494472.1 - 4 - 5 587 57
DQ494473.1 - 4 - 5 587 57
DQ494474.1 - 4 - 5 587 57
DQ494475.1 - 4 - 5 587 57
DQ494476.1 - 4 - 5 587 57
DQ494477.1 - 4 - 5 587 57
DQ494478.1 - 4 - 5 587 57
DQ494479.1 - 4 - 5 587 57
KJ534993.1 - 4 - 5 587 57
KJ535064.1 - 4 - 5 570 57

View Full Product Details