The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198994.1 | NP_001185923.1 | 5 - 6 | 785 | 101 |
XM_006710839.1 | XP_006710902.1 | 6 - 7 | 978 | 101 | |
GenBank mRNA | AF250320.1 | - | 5 - 6 | 785 | 101 |