The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024329.5 | NP_077305.2 | 3 - 4 | 669 | 74 |
GenBank mRNA | BC007233.1 | - | 3 - 4 | 627 | 74 |
BC014923.2 | - | 3 - 4 | 623 | 74 | |
BC023611.2 | - | 3 - 4 | 619 | 74 | |
BC068473.1 | - | 3 - 4 | 642 | 74 |