The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 10 - 11 | 1535 | 70 |
NM_001256879.1 | NP_001243808.1 | 11 - 12 | 1896 | 70 | |
NM_006230.3 | NP_006221.2 | 10 - 11 | 1362 | 70 | |
XM_011515431.1 | XP_011513733.1 | 10 - 11 | 1358 | 70 | |
GenBank mRNA | AK292347.1 | - | 11 - 12 | 1896 | 70 |
AK298168.1 | - | 10 - 11 | 1226 | 70 | |
AK312294.1 | - | 10 - 11 | 1495 | 70 | |
AW732014.1 | - | 1 - 2 | 70 | ||
BC000459.2 | - | 10 - 11 | 1325 | 70 | |
BC030001.2 | - | 9 - 10 | 1302 | 70 | |
U21090.1 | - | 10 - 11 | 1324 | 70 |