Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138734.2 NP_620063.1 1 - 2 779 89

View Full Product Details