Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP2
Entrez Gene ID
9099
Gene Name
ubiquitin specific peptidase 2
Gene Aliases
UBP41, USP9
Chromosome Location
Chr.11: 119355215 - 119381726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004205.4 NP_004196.4 2 - 3 1065 93
XM_005271721.4 XP_005271778.1 2 - 3 938 93
XM_005271722.2 XP_005271779.1 2 - 3 944 93
GenBank mRNA AK057225.1 - 2 - 3 994 93
BC002854.1 - 2 - 3 946 93
BC002955.1 - 2 - 3 946 93

View Full Product Details