The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006643.3 | NP_006634.3 | 2 - 3 | 427 | 128 |
GenBank mRNA | BC014515.1 | - | 2 - 3 | 294 | 128 |