The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004711.4 | NP_004702.2 | 2 - 3 | 404 | 66 |
NM_145731.3 | NP_663783.1 | 2 - 3 | 404 | 66 | |
NM_145738.2 | NP_663791.1 | 2 - 3 | 370 | 66 | |
GenBank mRNA | AJ002303.1 | - | 2 - 3 | 370 | 66 |
AJ002304.1 | - | 2 - 3 | 342 | 66 | |
AJ002305.1 | - | 2 - 3 | 342 | 66 | |
AK289507.1 | - | 2 - 3 | 351 | 66 | |
AK295863.1 | - | 2 - 3 | 362 | 66 | |
BC000731.2 | - | 2 - 3 | 368 | 66 | |
BI667990.1 | - | 2 - 3 | 428 | 66 | |
BT007135.1 | - | 2 - 3 | 342 | 66 | |
CR456590.1 | - | 2 - 3 | 352 | 66 |