Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
25878
Gene Name
matrix remodeling associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 3308565 - 3347098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015419.3 NP_056234.2 4 - 5 867 88
XM_005274485.2 XP_005274542.1 4 - 5 1322 88
GenBank mRNA AF245505.1 - 3 - 4 710 88
BC011846.2 - 4 - 5 864 88
BC064986.1 - 4 - 5 864 88
BC080586.1 - 4 - 5 864 88

View Full Product Details