The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022151.4 | NP_071434.2 | 1 - 2 | 97 | 141 |
GenBank mRNA | BG699993.1 | - | 1 - 2 | 106 | 141 |