The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015604.3 | NP_056419.2 | 9 - 10 | 1028 | 68 |
NM_181340.2 | NP_851937.1 | 8 - 9 | 928 | 68 | |
XM_017021207.1 | XP_016876696.1 | 9 - 10 | 1023 | 68 | |
XM_017021208.1 | XP_016876697.1 | 9 - 10 | 1023 | 68 | |
XM_017021210.1 | XP_016876699.1 | 9 - 10 | 1023 | 68 | |
XM_017021214.1 | XP_016876703.1 | 9 - 10 | 979 | 68 | |
XM_017021215.1 | XP_016876704.1 | 9 - 10 | 993 | 68 | |
GenBank mRNA | AK315499.1 | - | 9 - 10 | 1028 | 68 |
AL080157.1 | - | 8 - 9 | 799 | 68 | |
BC018979.2 | - | 9 - 10 | 840 | 68 | |
BC035901.1 | - | 8 - 9 | 782 | 68 | |
BX247962.1 | - | 9 - 10 | 855 | 68 |