Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX8
Entrez Gene ID
29886
Gene Name
sorting nexin 8
Gene Aliases
Mvp1
Chromosome Location
Chr.7: 2251770 - 2354481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013321.3 NP_037453.1 3 - 4 476 53
XM_011515329.2 XP_011513631.1 3 - 4 589 53
XM_011515330.1 XP_011513632.1 3 - 4 478 53
XM_017012083.1 XP_016867572.1 3 - 4 546 53
XM_017012084.1 XP_016867573.1 3 - 4 588 53
GenBank mRNA AF121858.1 - 3 - 4 432 53
AK074544.1 - 3 - 4 425 53
AK315670.1 - 3 - 4 446 53
BC007785.2 - 3 - 4 405 53
BC021565.1 - 3 - 4 465 53
BP320122.1 - 3 - 4 476 53

View Full Product Details