Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYG1
Entrez Gene ID
129530
Gene Name
lysozyme g1
Gene Aliases
SALW1939
Chromosome Location
Chr.2: 99284238 - 99304775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174898.2 NP_777558.1 5 - 6 579 62
XM_005263869.2 XP_005263926.1 6 - 7 679 62
XM_005263870.3 XP_005263927.1 6 - 7 584 62
XM_005263871.4 XP_005263928.1 6 - 7 715 62
XM_011510580.2 XP_011508882.1 6 - 7 678 62
XM_017003311.1 XP_016858800.1 4 - 5 845 62
XM_017003312.1 XP_016858801.1 3 - 4 896 62
XM_017003313.1 XP_016858802.1 4 - 5 563 62
GenBank mRNA AY358988.1 - 6 - 7 646 62
BC029126.1 - 5 - 6 579 62

View Full Product Details