The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006642.3 | NP_006633.1 | 6 - 7 | 847 | 99 |
XM_005273021.4 | XP_005273078.1 | 5 - 6 | 522 | 99 | |
XM_005273023.4 | XP_005273080.1 | 6 - 7 | 1524 | 99 | |
XM_006711727.3 | XP_006711790.1 | 6 - 7 | 733 | 99 | |
XM_017000105.1 | XP_016855594.1 | 6 - 7 | 834 | 99 | |
XM_017000106.1 | XP_016855595.1 | 6 - 7 | 815 | 99 | |
XM_017000107.1 | XP_016855596.1 | 5 - 6 | 522 | 99 | |
XM_017000109.1 | XP_016855598.1 | 6 - 7 | 733 | 99 | |
GenBank mRNA | AF161348.1 | - | 6 - 7 | 824 | 99 |
AF250731.1 | - | 6 - 7 | 678 | 99 | |
AK299212.1 | - | 6 - 7 | 796 | 99 | |
AK299249.1 | - | 3 - 4 | 439 | 99 | |
BC032454.1 | - | 6 - 7 | 790 | 99 |