The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152787.4 | NP_690000.3 | 9 - 10 | 2558 | 68 |
GenBank mRNA | AY331591.1 | - | 10 - 11 | 2442 | 68 |
AY371491.1 | - | 5 - 6 | 1895 | 68 | |
AY437560.1 | - | 5 - 6 | 1919 | 68 | |
BC032526.1 | - | 6 - 7 | 2096 | 68 |