The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021937.4 | NP_068756.2 | 2 - 3 | 571 | 143 |
XM_005247696.2 | XP_005247753.1 | 2 - 3 | 572 | 143 | |
XM_017006975.1 | XP_016862464.1 | 2 - 3 | 528 | 143 | |
XM_017006976.1 | XP_016862465.1 | 2 - 3 | 528 | 143 | |
XM_017006977.1 | XP_016862466.1 | 2 - 3 | 528 | 143 | |
XM_017006978.1 | XP_016862467.1 | 3 - 4 | 351 | 143 | |
XM_017006979.1 | XP_016862468.1 | 2 - 3 | 840 | 143 | |
XM_017006980.1 | XP_016862469.1 | 2 - 3 | 572 | 143 | |
GenBank mRNA | AF268872.1 | - | 2 - 3 | 310 | 143 |
BC007933.2 | - | 2 - 3 | 449 | 143 | |
BG420135.1 | - | 2 - 3 | 558 | 146 |