Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OXNAD1
Entrez Gene ID
92106
Gene Name
oxidoreductase NAD binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 16265160 - 16386979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138381.3 NP_612390.1 8 - 9 1289 100
XM_005265559.3 XP_005265616.1 8 - 9 1270 100
XM_006713405.1 XP_006713468.1 8 - 9 1253 100
XM_006713406.2 XP_006713469.1 9 - 10 1415 100
XM_011534230.2 XP_011532532.1 8 - 9 1270 100
XM_011534232.2 XP_011532534.1 8 - 9 1270 100
XM_011534233.2 XP_011532535.1 8 - 9 1270 100
XM_011534234.2 XP_011532536.1 8 - 9 1270 100
XM_011534235.2 XP_011532537.1 8 - 9 1270 100
XM_017007485.1 XP_016862974.1 8 - 9 1253 100
XM_017007486.1 XP_016862975.1 8 - 9 1285 100
XM_017007487.1 XP_016862976.1 8 - 9 1285 100
XM_017007488.1 XP_016862977.1 8 - 9 1285 100
XM_017007489.1 XP_016862978.1 8 - 9 1285 100
XM_017007490.1 XP_016862979.1 8 - 9 1285 100
XM_017007491.1 XP_016862980.1 8 - 9 1285 100
XM_017007492.1 XP_016862981.1 8 - 9 1024 100
XM_017007493.1 XP_016862982.1 9 - 10 1415 100
XM_017007494.1 XP_016862983.1 9 - 10 1415 100
XM_017007495.1 XP_016862984.1 9 - 10 1412 100
XM_017007496.1 XP_016862985.1 9 - 10 1412 100
GenBank mRNA AB209556.1 - 6 - 7 783 100
AK298829.1 - 7 - 8 844 100
AL832787.1 - 4 - 5 591 100
BC008322.1 - 8 - 9 1216 100
DQ364598.1 - 8 - 9 1247 100

View Full Product Details