Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNPC3
Entrez Gene ID
55599
Gene Name
RNA binding region (RNP1, RRM) containing 3
Gene Aliases
RBM40, RNP, SNRNP65
Chromosome Location
Chr.1: 103525956 - 103555239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017619.3 NP_060089.1 6 - 7 736 73
GenBank mRNA AK057799.1 - 6 - 7 715 73
AK289844.1 - 6 - 7 1001 73
AY099329.1 - 6 - 7 721 73
BC010697.1 - 6 - 7 712 73

View Full Product Details