Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO5
Entrez Gene ID
2330
Gene Name
flavin containing monooxygenase 5
Gene Aliases
-
Chromosome Location
Chr.1: 147183963 - 147225798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144830.2 NP_001138302.1 6 - 7 1507 134
NM_001461.3 NP_001452.2 8 - 9 1640 134
XM_005272946.4 XP_005273003.1 8 - 9 1614 134
XM_005272947.4 XP_005273004.1 8 - 9 1358 134
XM_005272948.4 XP_005273005.1 8 - 9 1341 134
XM_006711245.3 XP_006711308.1 7 - 8 1277 134
XM_011509350.2 XP_011507652.1 8 - 9 1645 134
XM_011509351.2 XP_011507653.1 7 - 8 1377 134
XM_017000801.1 XP_016856290.1 7 - 8 1389 134
XM_017000802.1 XP_016856291.1 7 - 8 1159 134
GenBank mRNA AK222728.1 - 8 - 9 1355 134
AK314647.1 - 8 - 9 1568 134
BC035687.1 - 6 - 7 1180 134
L37080.1 - 8 - 9 1326 134
Z47553.1 - 8 - 9 1332 134

View Full Product Details