The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282179.1 | NP_001269108.1 | 12 - 13 | 1792 | 87 |
NM_001282180.1 | NP_001269109.1 | 12 - 13 | 1795 | 87 | |
NM_001282181.1 | NP_001269110.1 | 11 - 12 | 1162 | 87 | |
NM_001282182.1 | NP_001269111.1 | 11 - 12 | 1117 | 87 | |
NM_006246.3 | NP_006237.1 | 12 - 13 | 1795 | 87 | |
NR_104104.1 | 11 - 12 | 1598 | 87 | ||
GenBank mRNA | AB209811.1 | - | 12 - 13 | 1748 | 87 |
AB451233.1 | - | 11 - 12 | 1205 | 87 | |
AB451358.1 | - | 11 - 12 | 1205 | 87 | |
AK299520.1 | - | 12 - 13 | 1732 | 87 | |
AK301796.1 | - | 11 - 12 | 1162 | 87 | |
AK313500.1 | - | 12 - 13 | 1647 | 87 | |
AK316430.1 | - | 11 - 12 | 1116 | 87 | |
BC013154.1 | - | 11 - 12 | 1591 | 87 | |
BC034994.1 | - | 13 - 14 | 1789 | 87 | |
BC048305.1 | - | 12 - 13 | 1528 | 87 | |
BC064358.1 | - | 12 - 13 | 1775 | 87 | |
BC073873.1 | - | 12 - 13 | 1747 | 87 | |
BC092477.1 | - | 12 - 13 | 1765 | 87 | |
BC093766.1 | - | 12 - 13 | 1263 | 87 | |
BC101479.1 | - | 12 - 13 | 1263 | 87 | |
BC143231.1 | - | 12 - 13 | 1260 | 87 | |
BC143234.1 | - | 12 - 13 | 1263 | 87 | |
L76703.1 | - | 12 - 13 | 1772 | 87 | |
Z69029.1 | - | 12 - 13 | 1710 | 87 |