Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 3 - 4 4024 128
NM_014866.1 NP_055681.1 4 - 5 4014 128
XM_005266134.2 XP_005266191.1 3 - 4 3877 128
XM_011519246.2 XP_011517548.1 4 - 5 4023 128
XM_011519247.1 XP_011517549.1 3 - 4 3880 128
XM_011519248.2 XP_011517550.1 3 - 4 3854 128
XM_011519249.1 XP_011517551.1 3 - 4 3877 128
XM_011519250.2 XP_011517552.1 3 - 4 3877 128
XM_011519251.1 XP_011517553.1 3 - 4 3877 128
XM_011519252.2 XP_011517554.1 3 - 4 3877 128
XM_011519253.1 XP_011517555.1 3 - 4 3877 128
XM_011519254.1 XP_011517556.1 3 - 4 3877 128
XM_011519255.2 XP_011517557.1 3 - 4 3877 128
XM_011519256.1 XP_011517558.1 3 - 4 3877 128
XM_011519257.1 XP_011517559.1 3 - 4 3877 128
XM_011519258.1 XP_011517560.1 3 - 4 3877 128
XM_011519259.1 XP_011517561.1 3 - 4 3877 128
XM_011519260.2 XP_011517562.1 3 - 4 3861 128
XM_011519261.1 XP_011517563.1 3 - 4 3877 128
XM_011519262.1 XP_011517564.1 4 - 5 3166 128
XM_011519263.2 XP_011517565.1 4 - 5 3166 128
XM_011519264.2 XP_011517566.1 3 - 4 3877 128
XM_017015363.1 XP_016870852.1 3 - 4 3877 128
XM_017015364.1 XP_016870853.1 3 - 4 3877 128
GenBank mRNA AB002308.4 - 4 - 5 4014 128
AB593115.1 - 3 - 4 4024 128
BC125018.1 - 2 - 3 2586 128
BC125019.1 - 3 - 4 1875 128
DQ903855.1 - 2 - 3 3171 128

View Full Product Details