The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015347.4 | NP_056162.4 | 13 - 14 | 2633 | 91 |
XM_011538102.1 | XP_011536404.1 | 15 - 16 | 3451 | 91 | |
XM_011538103.2 | XP_011536405.1 | 17 - 18 | 3913 | 91 | |
XM_011538104.2 | XP_011536406.1 | 17 - 18 | 3728 | 91 | |
XM_011538105.2 | XP_011536407.1 | 16 - 17 | 3185 | 91 | |
XM_011538106.2 | XP_011536408.1 | 15 - 16 | 3026 | 91 | |
XM_017019105.1 | XP_016874594.1 | 17 - 18 | 3926 | 91 | |
XM_017019106.1 | XP_016874595.1 | 16 - 17 | 3185 | 91 | |
XM_017019107.1 | XP_016874596.1 | 16 - 17 | 3007 | 91 | |
XM_017019108.1 | XP_016874597.1 | 16 - 17 | 3190 | 91 | |
XM_017019109.1 | XP_016874598.1 | 14 - 15 | 2734 | 91 | |
XM_017019110.1 | XP_016874599.1 | 16 - 17 | 3007 | 91 | |
XM_017019111.1 | XP_016874600.1 | 16 - 17 | 3007 | 91 | |
GenBank mRNA | AB002316.1 | - | 13 - 14 | 2633 | 91 |
AY699979.1 | - | 10 - 11 | 2324 | 91 |