Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HECW1
- Entrez Gene ID
-
23072
- Gene Name
- HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
- Gene Aliases
-
NEDL1
- Chromosome Location
-
Chr.7: 43112580 - 43566001 on Build GRCh38
View Full Product Details