The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103146.1 | NP_001096616.1 | 21 - 22 | 2725 | 59 |
NM_001103147.1 | NP_001096617.1 | 23 - 24 | 2858 | 59 | |
NM_001103148.1 | NP_001096618.1 | 20 - 21 | 2641 | 59 | |
NM_015575.3 | NP_056390.2 | 23 - 24 | 2865 | 59 | |
NR_103492.1 | 20 - 21 | 2641 | 59 | ||
GenBank mRNA | AB014542.2 | - | 23 - 24 | 2817 | 59 |
AK001739.1 | - | 23 - 24 | 2839 | 59 | |
AK302998.1 | - | 15 - 16 | 2227 | 59 | |
AY176045.1 | - | 23 - 24 | 2839 | 59 | |
BC014047.2 | - | 8 - 9 | 1049 | 59 | |
BC136251.1 | - | 20 - 21 | 2596 | 59 | |
BC143352.1 | - | 20 - 21 | 2596 | 59 | |
BC146775.1 | - | 23 - 24 | 2817 | 59 | |
BX537885.1 | - | 21 - 22 | 2660 | 59 | |
BX538172.1 | - | 20 - 21 | 2609 | 59 | |
BX538321.1 | - | 21 - 22 | 2681 | 59 | |
BX647172.1 | - | 5 - 6 | 1563 | 59 | |
BX647505.1 | - | 20 - 21 | 2472 | 59 |