Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OBSL1
Entrez Gene ID
23363
Gene Name
obscurin like 1
Gene Aliases
-
Chromosome Location
Chr.2: 219549408 - 219571573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246427.4 XP_005246484.1 9 - 10 3551 69
XM_011510866.2 XP_011509168.1 9 - 10 3554 69
XM_017003699.1 XP_016859188.1 9 - 10 3562 69
XM_017003700.1 XP_016859189.1 9 - 10 3560 69
GenBank mRNA AB014557.2 - 9 - 10 2731 69
AK299261.1 - 2 - 3 380 69

View Full Product Details