The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015311.2 | NP_056126.1 | 19 - 20 | 5725 | 94 |
XM_005246424.4 | XP_005246481.1 | 18 - 19 | 5473 | 94 | |
XM_011510857.2 | XP_011509159.1 | 19 - 20 | 5752 | 94 | |
XM_017003696.1 | XP_016859185.1 | 19 - 20 | 5751 | 94 | |
XM_017003697.1 | XP_016859186.1 | 19 - 20 | 5751 | 94 | |
XM_017003698.1 | XP_016859187.1 | 18 - 19 | 5474 | 94 | |
XM_017003699.1 | XP_016859188.1 | 18 - 19 | 5475 | 94 | |
XM_017003700.1 | XP_016859189.1 | 17 - 18 | 5197 | 94 | |
GenBank mRNA | AB014557.2 | - | 15 - 16 | 4310 | 94 |
EF063638.1 | - | 19 - 20 | 5468 | 94 |