The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006133.2 | NP_006124.1 | 7 - 8 | 891 | 109 |
XM_017018238.1 | XP_016873727.1 | 7 - 8 | 1140 | 109 | |
XM_017018239.1 | XP_016873728.1 | 7 - 8 | 1045 | 109 | |
XM_017018240.1 | XP_016873729.1 | 7 - 8 | 1140 | 109 | |
GenBank mRNA | AB014559.2 | - | 7 - 8 | 886 | 109 |
AY275377.1 | - | 6 - 7 | 775 | 109 | |
BC150176.1 | - | 7 - 8 | 891 | 109 | |
BC150195.1 | - | 7 - 8 | 891 | 109 | |
BC152453.1 | - | 7 - 8 | 886 | 109 |