The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318339.1 | NP_001305268.1 | 14 - 15 | 2661 | 74 |
NM_015308.3 | NP_056123.2 | 14 - 15 | 2655 | 74 | |
GenBank mRNA | AB023231.2 | - | 14 - 15 | 2654 | 74 |
AK022987.1 | - | 7 - 8 | 1099 | 74 | |
AK054636.1 | - | 14 - 15 | 2478 | 74 | |
AK095093.1 | - | 2 - 3 | 2449 | 74 | |
AL137480.1 | - | 14 - 15 | 2454 | 74 | |
BC037404.1 | - | 14 - 15 | 2486 | 74 | |
BX537864.1 | - | 2 - 3 | 2578 | 74 |