Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
55075
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
NUCLING
Chromosome Location
Chr.15: 70654554 - 70778823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008224.1 NP_001008225.1 2 - 3 537 70
NM_018003.2 NP_060473.2 2 - 3 314 70
XM_005254529.4 XP_005254586.1 2 - 3 564 70
XM_011521752.2 XP_011520054.2 2 - 3 643 70
XM_011521753.2 XP_011520055.2 2 - 3 644 70
XM_011521754.2 XP_011520056.1 2 - 3 274 70
XM_017022394.1 XP_016877883.1 2 - 3 396 70
XM_017022395.1 XP_016877884.1 2 - 3 174 70
GenBank mRNA AB046781.2 - 2 - 3 210 70
AF322916.1 - 2 - 3 259 70
AK055435.1 - 2 - 3 564 70
AL834243.1 - 2 - 3 539 70
BC016881.1 - 2 - 3 526 70
BC113407.1 - 2 - 3 233 70
BC113409.1 - 2 - 3 233 70
BC143266.1 - 2 - 3 233 70
BC143267.1 - 2 - 3 233 70
CN303461.1 - 2 - 3 315 70

View Full Product Details