Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
55075
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
NUCLING
Chromosome Location
Chr.15: 70654554 - 70778823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008224.1 NP_001008225.1 6 - 7 821 144
NM_018003.2 NP_060473.2 6 - 7 598 144
XM_005254529.4 XP_005254586.1 6 - 7 848 144
XM_011521752.2 XP_011520054.2 6 - 7 927 144
XM_011521753.2 XP_011520055.2 6 - 7 928 144
XM_011521754.2 XP_011520056.1 6 - 7 558 144
XM_017022394.1 XP_016877883.1 6 - 7 680 144
XM_017022395.1 XP_016877884.1 6 - 7 458 144
GenBank mRNA AB046781.2 - 6 - 7 494 144
AK055435.1 - 6 - 7 848 144
AL834243.1 - 6 - 7 823 144
BC016881.1 - 6 - 7 810 144
BC113407.1 - 6 - 7 517 144
BC113409.1 - 6 - 7 517 144
BC143266.1 - 6 - 7 517 144
CN303461.1 - 6 - 7 599 144

View Full Product Details