Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 16 - 17 2152 69
XM_005273013.4 XP_005273070.1 15 - 16 1984 69
XM_005273018.1 XP_005273075.1 15 - 16 1991 69
XM_005273021.4 XP_005273078.1 16 - 17 1924 69
XM_005273022.3 XP_005273079.1 13 - 14 1773 69
XM_006711727.3 XP_006711790.1 16 - 17 2038 69
XM_006711728.3 XP_006711791.1 15 - 16 1909 69
XM_011544021.2 XP_011542323.1 17 - 18 2134 69
XM_011544022.2 XP_011542324.1 17 - 18 2209 69
XM_011544026.2 XP_011542328.1 15 - 16 1897 69
XM_011544028.2 XP_011542330.1 13 - 14 1672 69
XM_011544030.2 XP_011542332.1 9 - 10 2685 69
XM_017000105.1 XP_016855594.1 14 - 15 1902 69
XM_017000106.1 XP_016855595.1 16 - 17 2120 69
XM_017000107.1 XP_016855596.1 17 - 18 2036 69
XM_017000108.1 XP_016855597.1 19 - 20 2583 69
GenBank mRNA AF039690.1 - 8 - 9 1086 69
AF250731.1 - 16 - 17 1983 69
AK297105.1 - 12 - 13 1571 69
AK299212.1 - 16 - 17 2101 69
AK299249.1 - 13 - 14 1744 69
BC045832.1 - 5 - 6 591 69

View Full Product Details